Learn More
1,1-Dibromoethane, 99%, stabilized, Thermo Scientific Chemicals
£103.00 - £429.00
Chemische Identifikatoren
CAS | 557-91-5 |
---|---|
Molecular Formula | C2H4Br2 |
Molecular Weight (g/mol) | 187.86 |
MDL Number | MFCD00041719 |
InChI Key | APQIUTYORBAGEZ-UHFFFAOYSA-N |
Synonym | ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552 |
PubChem CID | 11201 |
IUPAC Name | 1,1-dibromoethane |
SMILES | CC(Br)Br |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10147510
|
Thermo Scientific Acros
406480250 |
25 g | Glass bottle |
£103.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10470641
|
Thermo Scientific Acros
406481000 |
100 g | Glass bottle |
£429.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
557-91-5 | |
187.86 | |
APQIUTYORBAGEZ-UHFFFAOYSA-N | |
11201 | |
CC(Br)Br |
C2H4Br2 | |
MFCD00041719 | |
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552 | |
1,1-dibromoethane |
Spezifikation
557-91-5 | |
100.0 | |
2.0600g/mL | |
Authentic | |
C2H4Br2 | |
1.5090 to 1.5120 | |
2.06 | |
APQIUTYORBAGEZ-UHFFFAOYSA-N | |
1,1-dibromoethane | |
187.86 | |
187.86 | |
Liquid |
98.5 | |
Colorless to Brown | |
107.0°C | |
98.5% min. (GC) | |
CH3CHBr2 | |
MFCD00041719 | |
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552 | |
CC(Br)Br | |
potassium carbonate | |
11201 | |
99% | |
1,1-Dibromoethane, 99% |
Sicherheit und Handhabung
GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-184-9
missing translation for 'rtecsNumber' : KH9000000
missing translation for 'tsca' : TSCA
- 1,1-Dibromoethane, stabilized
Signalwort
- Warning
Gefahrenkategorie
- Acute toxicity Category 4
- Carcinogenicity Category 2
Gefahrenhinweis
- H351-Suspected of causing cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P313-Get medical advice/attention.
RUO – Research Use Only