missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 12229299
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 1115-70-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 218.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H12ClN5
MDL Number MFCD00012582
Quantity 5 g
Merck Index 15,601
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso (heated)
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N
IUPAC Name 3-(diaminomethylidene)-1,1-dimethylguanidine;hydrochloride
Molecular Weight (g/mol) 165.63
PubChem CID 14219
Formula Weight 165.63
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1, 1-Dimethylbiguanide hydrochloride
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 1,1-Dimethylbiguanide hydrochloride
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'