missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)ethylamine, 98%, Thermo Scientific Chemicals

Product Code. 15144805
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42882-31-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
InChI Key RTCUCQWIICFPOD-UHFFFAOYSA-N
Synonym +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine
PubChem CID 98089
IUPAC Name 1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Density 1.0630g/mL
Boiling Point 156°C (15.0 mmHg)
Flash Point >110°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.063
Solubility Information Solubility in water: insoluble
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material 1-(1-Naphthyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(1-Naphthyl)ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'