Learn More
1,10-Diaminodecane, 97%, Thermo Scientific Chemicals
£35.70 - £106.00
Chemical Identifiers
CAS | 646-25-3 |
---|---|
Molecular Formula | C10H24N2 |
Molecular Weight (g/mol) | 172.31 |
MDL Number | MFCD00008151 |
InChI Key | YQLZOAVZWJBZSY-UHFFFAOYSA-N |
Synonym | 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine |
PubChem CID | 1317 |
IUPAC Name | decane-1,10-diamine |
SMILES | C(CCCCCN)CCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10622115
|
Thermo Scientific Acros
112130050 |
5 g | Glass bottle |
£35.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540371
|
Thermo Scientific Acros
112130250 |
25 g | Glass bottle |
£106.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
646-25-3 | |
172.31 | |
YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
1317 | |
C(CCCCCN)CCCCN |
C10H24N2 | |
MFCD00008151 | |
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
decane-1,10-diamine |
Specifications
646-25-3 | |
100.0 | |
White to Yellow | |
Authentic | |
C10H24N2 | |
MFCD00008151 | |
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
C(CCCCCN)CCCCN | |
172.31 | |
172.31 | |
Chunks and/or Powder |
96.0 | |
62.0°C to 63.0°C | |
140.0°C (12.0 mmHg) | |
96% min. (GC) | |
H2N(CH2)10NH2 | |
04, 273 | |
YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
decane-1,10-diamine | |
1317 | |
97% | |
1, 10-Diaminodecane, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-471-9
missing translation for 'rtecsNumber' : HD7175000
missing translation for 'tsca' : TSCA
- 1,10-Diaminodecane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only