missing translation for 'onlineSavingsMsg'
Learn More
1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals Product Code.: 15427947

1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals

Product Code. 15427947
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15427947

Brand: Thermo Scientific Alfa Aesar H51063.MD

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23978-55-4
Molecular Formula C12H28N2O4
Molecular Weight (g/mol) 264.37
MDL Number MFCD00005112
InChI Key NLMDJJTUQPXZFG-UHFFFAOYSA-P
Synonym kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
PubChem CID 72805
IUPAC Name 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
SMILES C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

Specifications

Quantity 250 mg
Beilstein 609764
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 262.35
Percent Purity 96%
Chemical Name or Material 1,10-Diaza-18-crown-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.