missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane, 97%, Thermo Scientific Chemicals

Product Code. 11429493
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr

Specifications

Melting Point 26°C to 30°C
Density 1.34
Boiling Point 157°C to 159°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.493
Quantity 500 g
Beilstein 506156
Solubility Information Insoluble in water.
Formula Weight 300.08
Percent Purity 97%
Chemical Name or Material 1,10-Dibromodecane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'