Learn More
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
£44.12 - £311.55
Chemical Identifiers
CAS | 4101-68-2 |
---|---|
Molecular Formula | C10H20Br2 |
Molecular Weight (g/mol) | 300.08 |
MDL Number | MFCD00000222 |
InChI Key | GTQHJCOHNAFHRE-UHFFFAOYSA-N |
Synonym | decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromoShow More |
PubChem CID | 221483 |
IUPAC Name | 1,10-dibromodecane |
SMILES | BrCCCCCCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10386220
|
Thermo Scientific
112780250 |
25 mL | Glass bottle |
£44.12
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10132810
|
Thermo Scientific Acros
112781000 |
100 mL | Glass bottle |
£90.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617763
|
Thermo Scientific Acros
112785000 |
500 mL | Glass bottle |
£311.55
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4101-68-2 | |
300.08 | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
221483 | |
BrCCCCCCCCCCBr |
C10H20Br2 | |
MFCD00000222 | |
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromoShow More | |
1,10-dibromodecane |
Specifications
4101-68-2 | |
100.0 | |
Beige to Brown | |
160°C (15.0 mmHg) | |
Authentic | |
C10H20Br2 | |
1.4912 | |
01, I, 64 | |
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
1,10-dibromodecane | |
221483 | |
97% | |
1, 10-Dibromodecane |
96.0 | |
25°C to 30°C | |
1.3400g/mL | |
>112°C | |
96% min. (GC) | |
Br(CH2)10Br | |
MFCD00000222 | |
1.34 | |
Solubility in water: insoluble | |
BrCCCCCCCCCCBr | |
300.08 | |
300.07 | |
Crystalline Low Melting Solid |
Safety and Handling
missing translation for 'einecsNumber' : 223-871-
Recommended Storage : Product may darken during storage
- 1,10-Dibromodecane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.