missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
Description
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, bases and reducing agents.
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, bases and reducing agents.
Specifications
Specifications
| Quantity | 1 g |
| IUPAC Name | 1,10-phenanthroline-2,9-dicarboxylate |
| Formula Weight | 268.23 (Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | 1,10-Phenanthroline-2,9-dicarboxylic acid hydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction