missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals

Catalog Number 10695972
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
66-71-7
C12H8N2
180.21
MFCD00011678
DGEZNRSVGBDHLK-UHFFFAOYSA-N
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
1318
CHEBI:44975
1,10-phenanthroline
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1
66-71-7
C12H8N2
180.21
MFCD00011678
DGEZNRSVGBDHLK-UHFFFAOYSA-N
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
1318
CHEBI:44975
1,10-phenanthroline
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 117.0°C to 120.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 23,227
Merck Index 15,7328
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
Formula Weight 180.21
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 1, 10-Phenanthroline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'