Learn More
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Description
1,10-Phenanthroline iron(II) perchlorate is used as an oxidation, reduction indicator. It is used for the detection, identification, analysis, of chemical, biological, or pathologic processes or conditions. Agents that are capable of inserting themselves between the successive bases in DNA, thus kinking, uncoiling or otherwise deforming it and therefore preventing its proper functioning. They are used in the study of DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Red |
Quantity | 10 g |
UN Number | UN1479 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 795.37 |
Physical Form | Crystalline |
Chemical Name or Material | 1,10-Phenanthroline iron(II) perchlorate |
RUO – Research Use Only