missing translation for 'onlineSavingsMsg'
Learn More
1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals Product Code.: 10577172

1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 10577172
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10577172

Brand: Thermo Scientific Acros 417120010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-89-8, 66-71-7
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 97°C to 101°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Beilstein 23, 227
Merck Index 15, 7328
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,acetone and benzene
IUPAC Name 1,10-phenanthroline;hydrate
Formula Weight 198.23
Grade ACS Reagent
Physical Form Needle-like Crystals
Chemical Name or Material 1, 10-Phenanthroline monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0428021 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number417120010,417121000,417120250,417120050
Lot NumberA0444793 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number417120010,417121000,417120250,417120050
Lot NumberA0440086 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number417120010,417121000,417120250,417120050
Lot NumberA0436782 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number417120010,417121000,417120250,417120050
Lot NumberA0401297 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number417120010,417121000,417120250,417120050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.