missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Monohydrate, Certified AR for Analysis, Fisher Chemical™

£40.55 - £955.00

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline;hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10498410
View Documents
Fisher Chemical
P/2200/43
1 g Vial
£40.55
1g
Please to purchase this item. Need a web account? Register with us today!
10171910
View Documents Promotion Details
Fisher Chemical
P/2200/44
5 g Amber glass bottle
£158.00
5g
Please to purchase this item. Need a web account? Register with us today!
11498952
View Documents
Fisher Chemical
P/2200/48
100 g Plastic powder jar
£955.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5144-89-8
198.23
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
C12H10N2O
MFCD00149973
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
1,10-phenanthroline;hydrate

Specifications

5144-89-8
White
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline;hydrate
21226
Powder
97°C
C12H10N2O
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
198.23
198.23g/mol
Videos
SDS
Documents

Documents

Product Certifications