missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼225°C (decomposition) |
Odor | Odorless |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 4007967 |
Solubility Information | Soluble in water. |
Formula Weight | 234.68 (216.66 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | 1,10-Phenanthroline monohydrochloride monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'