missing translation for 'onlineSavingsMsg'
Learn More

1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals

Product Code. 11316866
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2783-17-7
Molecular Formula C12H28N2
Molecular Weight (g/mol) 200.37
MDL Number MFCD00008155
InChI Key QFTYSVGGYOXFRQ-UHFFFAOYSA-N
Synonym 1,12-diaminododecane, 1,12-dodecanediamine, dodecamethylenediamine, dodecyldiamine, 1,12-dodecamethylenediamine, 1,12-diamindodecane, 1,12'-dodecylenediamine, 1,12'-dodecamethylenediamine, unii-j3lm80w9nt, 1,12-dodecylenediamine
PubChem CID 17727
ChEBI CHEBI:49385
IUPAC Name dodecane-1,12-diamine
SMILES C(CCCCCCN)CCCCCN

Specifications

Melting Point 68°C to 72°C
Flash Point 155°C (311°F)
Linear Formula H2N(CH2)12NH2
Quantity 5 g
UN Number UN3259
Beilstein 1742765
Solubility Information Soluble in ethanol.
Formula Weight 200.37
Percent Purity ≥98%
Chemical Name or Material 1,12-Diaminododecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'