missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dibromododecane, 96%, Thermo Scientific Chemicals
£18.18 - £566.00
Chemical Identifiers
CAS | 3344-70-5 |
---|---|
Molecular Formula | C12H24Br2 |
Molecular Weight (g/mol) | 328.13 |
MDL Number | MFCD00000226 |
InChI Key | ZJJATABWMGVVRZ-UHFFFAOYSA-N |
Synonym | dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h |
PubChem CID | 18766 |
IUPAC Name | 1,12-dibromododecane |
SMILES | C(CCCCCCBr)CCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10591174
|
Thermo Scientific Acros
158450050 |
5 g | Glass bottle | ||||||
10367380
|
Thermo Scientific Acros
158450250 |
25 g | Glass bottle |
£66.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10486810
|
Thermo Scientific Acros
158451000 |
100 g | Glass bottle |
£185.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10207163
|
Thermo Scientific Acros
158455000 |
500 g | Glass bottle |
£566.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3344-70-5 | |
328.13 | |
ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
18766 | |
C(CCCCCCBr)CCCCCBr |
C12H24Br2 | |
MFCD00000226 | |
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
1,12-dibromododecane |
Specifications
3344-70-5 | |
100.0 | |
White to Brown | |
>110°C | |
96% | |
Br(CH2)12Br | |
01, II, 134 | |
ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
1,12-dibromododecane | |
18766 | |
96% | |
1,12-Dibromododecane |
95.0 | |
37.0°C to 42.0°C | |
215.0°C (15.0 mmHg) | |
Authentic | |
C12H24Br2 | |
MFCD00000226 | |
dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
C(CCCCCCBr)CCCCCBr | |
328.13 | |
328.13 | |
Crystalline Solid |
Safety and Handling
missing translation for 'einecsNumber' : 222-096-5
RUO – Research Use Only