missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,18-Octadecanedicarboxylic acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Description
1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 127°C |
Linear Formula | HO2C(CH2)18CO2H |
Quantity | 250 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 342.52 |
Physical Form | Powder |
Chemical Name or Material | 1,18-Octadecanedicarboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'