missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,18-Octadecanedicarboxylic acid, Thermo Scientific Chemicals
£29.40 - £1617.00
Chemical Identifiers
CAS | 2424-92-2 |
---|---|
Molecular Formula | C20H38O4 |
Molecular Weight (g/mol) | 342.52 |
MDL Number | MFCD00059627 |
InChI Key | JJOJFIHJIRWASH-UHFFFAOYSA-N |
Synonym | eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid |
PubChem CID | 75502 |
ChEBI | CHEBI:73728 |
IUPAC Name | icosanedioic acid |
SMILES | C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375017
|
Thermo Scientific Alfa Aesar
042004.04 |
2 g |
£29.40
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355017
|
Thermo Scientific Alfa Aesar
042004.09 |
10 g |
£115.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365017
|
Thermo Scientific Alfa Aesar
042004.18 |
50 g |
£439.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345017
|
Thermo Scientific Alfa Aesar
042004.30 |
250 g |
£1617.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2424-92-2 | |
342.52 | |
JJOJFIHJIRWASH-UHFFFAOYSA-N | |
75502 | |
icosanedioic acid |
C20H38O4 | |
MFCD00059627 | |
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid | |
CHEBI:73728 | |
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O |
Specifications
2424-92-2 | |
C20H38O4 | |
MFCD00059627 | |
Slightly soluble in water. | |
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O | |
342.52 | |
CHEBI:73728 | |
Powder |
127°C | |
HO2C(CH2)18CO2H | |
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid | |
JJOJFIHJIRWASH-UHFFFAOYSA-N | |
icosanedioic acid | |
75502 | |
342.52 | |
1,18-Octadecanedicarboxylic acid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only