missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[2-(2,5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl piperazine, 1-2-2,5-dimethyl-pyrrol-1-yl-ethyl-piperazine, 1-2-2,5-dimethylpyrrol-1-yl ethyl piperazine, piperazine,1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl, 2-2,5-dimethylpyrrolyl ethyl piperazine, enamine_004518, piperazine, 1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl |
Formula Weight | 207.32 |
Percent Purity | 97% |
Chemical Name or Material | 1-[2-(2, 5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'