missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5,6-Hexabromocyclohexane, Thermo Scientific Chemicals
Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation
£83.00 - £305.00
Chemical Identifiers
CAS | 1837-91-8 |
---|---|
Molecular Formula | C6H6Br6 |
Molecular Weight (g/mol) | 557.54 |
MDL Number | MFCD00059127 |
InChI Key | QFQZKISCBJKVHI-UHFFFAOYSA-N |
Synonym | benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc |
PubChem CID | 74603 |
IUPAC Name | 1,2,3,4,5,6-hexabromocyclohexane |
SMILES | BrC1C(Br)C(Br)C(Br)C(Br)C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453559
|
Thermo Scientific Alfa Aesar
J60853.03 |
1 g |
£83.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15463559
|
Thermo Scientific Alfa Aesar
J60853.06 |
5 g | N/A | ||||||
Description
Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1837-91-8 | |
557.54 | |
QFQZKISCBJKVHI-UHFFFAOYSA-N | |
74603 | |
BrC1C(Br)C(Br)C(Br)C(Br)C1Br |
C6H6Br6 | |
MFCD00059127 | |
benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc | |
1,2,3,4,5,6-hexabromocyclohexane |
Specifications
1837-91-8 | |
C6H6Br6 | |
benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc | |
BrC1C(Br)C(Br)C(Br)C(Br)C1Br | |
557.54 | |
557.5 | |
1,2,3,4,5,6-Hexabromocyclohexane |
White | |
MFCD00059127 | |
QFQZKISCBJKVHI-UHFFFAOYSA-N | |
1,2,3,4,5,6-hexabromocyclohexane | |
74603 | |
Solid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only