Learn More
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, CTC-Q-PHOS, 95%, Thermo Scientific Chemicals
Description
Heck and Suzuki reactions1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene is a sterically hindered phosphine ligand, which is used in palladium-catalyzed amination, arylation and etherification reactions of aryl chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
Quantity | 0.1 g |
Solubility Information | Soluble in ether,benzene,toluene and terahydrofuran. |
Formula Weight | 710.72 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 1,2,3,4,5-Pentaphenyl-1'- (di-tert-butylphosphino)ferrocene, CTC-Q-PHOS |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.