missing translation for 'onlineSavingsMsg'
Learn More
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals Product Code.: 10071550

1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals

Product Code. 10071550
1 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10071550

Brand: Thermo Scientific Acros 146730010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -35.0°C
Density 0.9730g/mL
Boiling Point 207.0°C
Flash Point 77°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C10H12
Refractive Index 1.5400 to 1.5420
Quantity 1 L
Beilstein 05, IV, 1388
Fieser 01,1142
Merck Index 15, 9368
Specific Gravity 0.973
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Viscosity 2.2 mPa.s (20°C)
Formula Weight 132.2
Percent Purity 98+%
Grade Pure
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,3,4-Tetrahydronaphthalene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : 1,2,3,4-Tetrahydronaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.