missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydronaphthalene, Reagent Grade, 99%, Honeywell™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
1 L
4 L
Packaging:
Glass Bottle
Specifications
Melting Point | -35°C |
Color | Colorless |
Density | 0.973g/mL at 25°C |
Boiling Point | 207°C |
Flash Point | 71°C (159.8°F) |
Infrared Spectrum | Conforms to structure |
Packaging | Glass Bottle |
Refractive Index | n20/D 1.541 |
Quantity | 100 mL |
UN Number | UN3082 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'