Learn More
1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
£17.10 - £402.00
Chemical Identifiers
CAS | 635-46-1 |
---|---|
Molecular Formula | C9H11N |
Molecular Weight (g/mol) | 133.19 |
MDL Number | MFCD00006693 |
InChI Key | LBUJPTNKIBCYBY-UHFFFAOYSA-N |
Synonym | tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline |
PubChem CID | 69460 |
ChEBI | CHEBI:213323 |
IUPAC Name | 1,2,3,4-tetrahydroquinoline |
SMILES | C1CNC2=CC=CC=C2C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10765801
|
Thermo Scientific Acros
138270250 |
25 g | Glass bottle |
£17.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503881
|
Thermo Scientific Acros
138271000 |
100 g | Glass bottle |
£36.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10675762
|
Thermo Scientific Acros
138275000 |
500 g | Glass bottle |
£107.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996491
|
Thermo Scientific Acros
138270025 |
2.5 kg | Glass bottle |
£402.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
635-46-1 | |
133.19 | |
LBUJPTNKIBCYBY-UHFFFAOYSA-N | |
69460 | |
1,2,3,4-tetrahydroquinoline |
C9H11N | |
MFCD00006693 | |
tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline | |
CHEBI:213323 | |
C1CNC2=CC=CC=C2C1 |
Specifications
635-46-1 | |
Yellow | |
249.0°C | |
Authentic | |
C9H11N | |
MFCD00006693 | |
1.06 | |
Solubility in water: <1g/L (20°C). Other solubilities: soluble in ethanol | |
C1CNC2=CC=CC=C2C1 | |
133.19 | |
CHEBI:213323 | |
133.19 | |
Liquid |
15.0°C to 17.0°C | |
1.0600g/mL | |
100°C | |
97.5% min. (GC) | |
1.5930 to 1.5950 | |
20,262 | |
tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline | |
LBUJPTNKIBCYBY-UHFFFAOYSA-N | |
1,2,3,4-tetrahydroquinoline | |
69460 | |
12.5 mPa.s (20°C) | |
98% | |
1,2,3,4-Tetrahydroquinoline |
Safety and Handling
GHS H Statement
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-237-6
Recommended Storage : Product may darken during storage
- 1,2,3,4-Tetrahydroquinoline
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only