missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetramethylbenzene, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.9 |
Boiling Point | 203°C |
Flash Point | 68°C (154°F) |
Refractive Index | 1.519 |
Quantity | 5 g |
Beilstein | 1904390 |
Solubility Information | Immiscible with water. |
Formula Weight | 134.22 |
Percent Purity | 95% |
Chemical Name or Material | 1,2,3,4-Tetramethylbenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'