missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydropyridine, 98%, Thermo Scientific Chemicals

Product Code. 10654935
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-05-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00005949
InChI Key FTAHXMZRJCZXDL-UHFFFAOYSA-N
Synonym pyridine, 1,2,3,6-tetrahydro, 3-piperideine, 1,2,5,6-tetrahydropyridine, unii-26rls9d255, delta 3-piperideine, .delta.3-piperidine, delta3-piperidine, acmc-1bcyc, 1,3,6-tetrahydropyridine, 1,5,6-tetrahydropyridine
PubChem CID 12750
ChEBI CHEBI:47860
IUPAC Name 1,2,3,6-tetrahydropyridine
SMILES C1CNCC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -48.0°C
Color Cream-Yellow to White
Density 0.9100g/mL
Boiling Point 108.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.48
Quantity 5 g
Beilstein 20, III, 1912
Specific Gravity 0.91
Solubility Information Solubility in water: soluble.
Formula Weight 83.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,6-Tetrahydropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,3,6-Tetrahydropyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'