missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dihydrobenzo[b]furan-5-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10191072
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 90843-31-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00068033
InChI Key MMVUJVASBDVNGJ-UHFFFAOYSA-N
Synonym 5-acetyl-2,3-dihydrobenzo b furan, 5-acetyl-2,3-dihydrobenzofuran, 1-2,3-dihydro-1-benzofuran-5-yl ethanone, 1-2,3-dihydrobenzofuran-5-yl ethanone, 1-2,3-dihydrobenzo b furan-5-yl ethan-1-one, 1-2,3-dihydro-benzofuran-5-yl-ethanone, 1-2,3-dihydro-5-benzofuranyl ethanone, 1-2,3-dihydro-1-benzofuran-5-yl ethan-1-one, 5-acetyl-2,3-dihydro-1-benzofuran
PubChem CID 145220
IUPAC Name 1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
SMILES CC(=O)C1=CC=C2OCCC2=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1-(2, 3-Dihydrobenzo[b]furan-5-yl)ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'