missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrabromobenzene, 94%, Thermo Scientific Chemicals

Product Code. 11303137
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-28-2
Molecular Formula C6H2Br4
Molecular Weight (g/mol) 393.70
MDL Number MFCD00000063
InChI Key QCKHVNQHBOGZER-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo
PubChem CID 12486
IUPAC Name 1,2,4,5-tetrabromobenzene
SMILES BrC1=CC(Br)=C(Br)C=C1Br

Specifications

Melting Point 175°C to 180°C
Quantity 50 g
Beilstein 1365830
Solubility Information Insoluble in water.
Formula Weight 393.7
Percent Purity 94%
Chemical Name or Material 1,2,4,5-Tetrabromobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'