missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4,5-Tetramethylbenzene, 97+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 78.0°C to 82.0°C |
| Boiling Point | 196.0°C to 197.0°C |
| Flash Point | 73°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% min. (GC) |
| Linear Formula | C6H2(CH3)4 |
| Quantity | 100 g |
| Beilstein | 05,431 |
| Merck Index | 15,3515 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene |
| Show More |
Safety and Handling
Product Identifier
- 1,2,4,5-Tetramethylbenzene
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Flammable solid Category 1
- H228-Flammable solid.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : 1,2,4,5-Tetramethylbenzene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction