missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylbenzene, 97+%

Product Code. 11361607
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11361607

Brand: Thermo Scientific Alfa Aesar L04493.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-93-2 | C10H14 | 134.222 g/mol

1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

Solubility
Soluble in alcohol, ether, benzene, and acetone. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008528
InChI Key SQNZJJAZBFDUTD-UHFFFAOYSA-N
Synonym durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
PubChem CID 7269
ChEBI CHEBI:38978
IUPAC Name 1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C=C1C)C)C

Specifications

Melting Point 77°C to 80°C
Boiling Point 191°C to 193°C
Flash Point 73°C (163°F)
Quantity 500 g
UN Number UN1325
Beilstein 1903393
Merck Index 14,3468
Solubility Information Soluble in alcohol,ether,benzene,and acetone. Insoluble in water.
Formula Weight 134.22
Percent Purity ≥97%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.