Learn More
1,2,4-Butanetriol Standard Solution For GC ASTM Method D-6584, Chem Lab
Brand: Chem Lab CL40.0272.0005
Additional Details : Weight : 0.00500kg Transport : UN number : 1282 Chem class : 3 Pack group : II
Specifications
Contains 1000 μg/mL in pyridine | |
5 mL | |
Standard | |
79.1 | |
GC ASTM Method D-6584 | |
Pyridine | |
115°C | |
Liquid |
Ampoule in glass | |
Colorless | |
1282 | |
1000 μg/mL in pyridine | |
0.98 g/mL | |
-42°C | |
17 °C |
Safety and Handling
- 1,2,4-Butanetriol standard solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
- MIXTURE LIST-Contains : 1,2,4-butantriol