missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4-Dimethylphenyl)ethanol, 95%

Product Code. 11360066
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11360066

Brand: Thermo Scientific Alfa Aesar H32141.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5379-19-1 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5379-19-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00060890
InChI Key DNHQUGRUHBFDFT-UHFFFAOYSA-N
Synonym 1-2,4-dimethylphenyl ethanol, 1-2,4-dimethylphenyl ethan-1-ol, 1-2,4-dimethylbenzene-1-ethanol
PubChem CID 21475
IUPAC Name 1-(2,4-dimethylphenyl)ethanol
SMILES CC1=CC(=C(C=C1)C(C)O)C

Specifications

Quantity 10 g
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 1-(2,4-Dimethylphenyl)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.