missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,5-Oxadiazole-3-carboxylic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 114.06 |
Percent Purity | 97% |
Chemical Name or Material | 1, 2, 5-Oxadiazole-3-carboxylic acid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 1,2,5-Oxadiazole-3-carboxylic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- MIXTURE LIST-Contains : 1,2,5-Oxadiazole-3-carboxylic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'