Learn More
1-(2-Aminoethyl)-4-methylpiperazine, 97+%, Thermo Scientific Chemicals
£37.79 - £442.00
Chemical Identifiers
CAS | 934-98-5 |
---|---|
Molecular Formula | C7H17N3 |
Molecular Weight (g/mol) | 143.234 |
MDL Number | MFCD03701701 |
InChI Key | GOWUDHPKGOIDIX-UHFFFAOYSA-N |
Synonym | 2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine |
PubChem CID | 70284 |
ChEBI | CHEBI:84290 |
IUPAC Name | 2-(4-methylpiperazin-1-yl)ethanamine |
SMILES | CN1CCN(CC1)CCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15469057
|
Thermo Scientific Alfa Aesar
H52371.MD |
250 mg |
£37.79
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15449057
|
Thermo Scientific Alfa Aesar
H52371.03 |
1 g |
£103.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15459057
|
Thermo Scientific Alfa Aesar
H52371.06 |
5 g |
£442.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Chemical Identifiers
934-98-5 | |
143.234 | |
GOWUDHPKGOIDIX-UHFFFAOYSA-N | |
70284 | |
2-(4-methylpiperazin-1-yl)ethanamine |
C7H17N3 | |
MFCD03701701 | |
2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine | |
CHEBI:84290 | |
CN1CCN(CC1)CCN |
Specifications
934-98-5 | |
MFCD03701701 | |
Air Sensitive | |
GOWUDHPKGOIDIX-UHFFFAOYSA-N | |
2-(4-methylpiperazin-1-yl)ethanamine | |
70284 | |
143.43 | |
1-(2-Aminoethyl)-4-methylpiperazine |
C7H17N3 | |
UN2735 | |
2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine | |
CN1CCN(CC1)CCN | |
143.234 | |
CHEBI:84290 | |
≥97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H312-H314-H317-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 213-296-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only