missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)imidazolidin-2-one , Tech., Thermo Scientific™

Product Code. 10745784
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6281-42-1
Molecular Formula C5H11N3O
Molecular Weight (g/mol) 129.163
MDL Number MFCD00086348
InChI Key PODSUMUEKRUDEI-UHFFFAOYSA-N
Synonym 1-2-aminoethyl imidazolidin-2-one, 1-2-aminoethyl-2-imidazolidone, 1-2-aminoethyl-2-imidazolidinone, 2-imidazolidinone, 1-2-aminoethyl, unii-9hkk1mly77, 1-2-aminoethyl imidazolidinone, 1-beta-aminoethyl-2-imidazolidone, 9hkk1mly77, n-2-aminoethyl ethyleneurea, aminoethyl imidazolidone
PubChem CID 80480
IUPAC Name 1-(2-aminoethyl)imidazolidin-2-one
SMILES C1CN(C(=O)N1)CCN

Specifications

Packaging Amber glass bottle
Quantity 10 g
Formula Weight 129.16
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(2-Aminoethyl)imidazolidin-2-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'