missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Aminophenyl)pyrrole, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1-(2-Aminophenyl)pyrrole is used in the synthesis of 4-substituted pyrrolo[1,2-a]quinoxaline derivatives. It participates in Pt(IV)-catalyzed hydroamination triggered cyclization reaction to yield fused pyrrolo [1,2-a] quinoxalines. It is also used as primary and secondary intermediates. They act as antileishmanial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Aminophenyl)pyrrole is used in the synthesis of 4-substituted pyrrolo[1,2-a]quinoxaline derivatives. It participates in Pt(IV)-catalyzed hydroamination triggered cyclization reaction to yield fused pyrrolo [1,2-a] quinoxalines. It is also used as primary and secondary intermediates. They act as antileishmanial agents.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides, Chloroformates. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
1-(2-Aminophenyl)pyrrole is used in the synthesis of 4-substituted pyrrolo[1,2-a]quinoxaline derivatives. It participates in Pt(IV)-catalyzed hydroamination triggered cyclization reaction to yield fused pyrrolo [1,2-a] quinoxalines. It is also used as primary and secondary intermediates. They act as antileishmanial agents.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides, Chloroformates. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 93°C to 94°C |
Quantity | 5 g |
Beilstein | 1307631 |
Solubility Information | Insoluble in water. |
Formula Weight | 158.2 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-(2-Aminophenyl)pyrrole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'