missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisothiazol-3-one, 97%

Product Code. 15476958
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15476958

Brand: Thermo Scientific Alfa Aesar H60492.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2634-33-5 | C7H5NOS | 151.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is an antimicrobial agent and a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide, methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2634-33-5
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00127753
InChI Key DMSMPAJRVJJAGA-UHFFFAOYSA-N
Synonym 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
PubChem CID 17520
ChEBI CHEBI:167099
IUPAC Name 1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2

Specifications

Melting Point 154°C to 158°C
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide,methanol.
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 1,2-Benzisothiazol-3-one
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Benzisothiazol-3-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Benzisothiazol-3(2H)-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.