missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzisoxazole, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water. 4.1 mg/mL in water at 25°C
Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
Soluble in water. 4.1 mg/mL in water at 25°C
Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.169 |
Boiling Point | 90°C to 92°C (15 mmHg) |
Flash Point | 86°C (186°F) |
Refractive Index | 1.562 |
Quantity | 5 g |
Beilstein | 2154 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. 4.1mg/mL in water at 25°C |
Formula Weight | 119.12 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'