missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-nitrophenoxy)ethane, 98%, Thermo Scientific™

Product Code. 11480277
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51661-19-9
Molecular Formula C14H12N2O6
Molecular Weight (g/mol) 304.26
MDL Number MFCD00024226
InChI Key DEHTVRKGDUCXRF-UHFFFAOYSA-N
Synonym 1,2-bis o-nitrophenoxy ethane, 1,2-bis 2-nitrophenoxy ethane, 1-nitro-2-2-2-nitrophenoxy ethoxy benzene, ethylene glycol bis 2-nitrophenyl ether, 1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene, bis 2-nitrophenoxy ethane, acmc-20a59z, 1,2-di o-nitrophenoxy ethane, 2,2'-ethylenedioxy bis 1-nitrobenzene, 2-nitro-1-2-2-nitrophenoxy ethoxy benzene
PubChem CID 394580
IUPAC Name 1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1OCCOC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 167°C to 170°C
Quantity 50 g
Formula Weight 304.26
Percent Purity 98%
Chemical Name or Material 1,2-Bis(2-nitrophenoxy)ethane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'