missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[2-(trifluoromethyl)phenyl]ethane, 97+%, Thermo Scientific™

£17.38 - £65.20

Chemical Identifiers

CAS 287172-66-1
Molecular Formula C16H12F6
Molecular Weight (g/mol) 318.262
MDL Number MFCD01631639
InChI Key OBMDBKNYJGWWJX-UHFFFAOYSA-N
Synonym 1,2-bis 2'-trifluoromethyl phenyl ethane, 1-trifluoromethyl-2-2-2-trifluoromethyl phenyl ethyl benzene, 1,2-bis-2-trifluoromethylphenyl ethane, 1,2-bis-2-trifluoromethyl phenyl ethane, 1,2-bis 2 trifluoromethyl phenyl ethane
PubChem CID 2773232
IUPAC Name 1-(trifluoromethyl)-2-[2-[2-(trifluoromethyl)phenyl]ethyl]benzene
SMILES C1=CC=C(C(=C1)CCC2=CC=CC=C2C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440368
View Documents
Thermo Scientific Alfa Aesar
B24533.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11450368
View Documents
Thermo Scientific Alfa Aesar
B24533.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

287172-66-1
318.262
OBMDBKNYJGWWJX-UHFFFAOYSA-N
2773232
C1=CC=C(C(=C1)CCC2=CC=CC=C2C(F)(F)F)C(F)(F)F
C16H12F6
MFCD01631639
1,2-bis 2'-trifluoromethyl phenyl ethane, 1-trifluoromethyl-2-2-2-trifluoromethyl phenyl ethyl benzene, 1,2-bis-2-trifluoromethylphenyl ethane, 1,2-bis-2-trifluoromethyl phenyl ethane, 1,2-bis 2 trifluoromethyl phenyl ethane
1-(trifluoromethyl)-2-[2-[2-(trifluoromethyl)phenyl]ethyl]benzene

Specifications

287172-66-1
C16H12F6
1,2-bis 2'-trifluoromethyl phenyl ethane, 1-trifluoromethyl-2-2-2-trifluoromethyl phenyl ethyl benzene, 1,2-bis-2-trifluoromethylphenyl ethane, 1,2-bis-2-trifluoromethyl phenyl ethane, 1,2-bis 2 trifluoromethyl phenyl ethane
C1=CC=C(C(=C1)CCC2=CC=CC=C2C(F)(F)F)C(F)(F)F
318.262
318.26
1,2-Bis[2-(trifluoromethyl)phenyl]ethane
74°C to 76°C
MFCD01631639
OBMDBKNYJGWWJX-UHFFFAOYSA-N
1-(trifluoromethyl)-2-[2-[2-(trifluoromethyl)phenyl]ethyl]benzene
2773232
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications