missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™ Product Code.: 10331150

1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™

Product Code. 10331150
250 mg
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy

Product Code. 10331150

Brand: Thermo Scientific Acros 435202500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 569650-64-2
Molecular Formula C34H56BF4P2Rh
Molecular Weight (g/mol) 716.48
MDL Number MFCD07369039
InChI Key AXLKBYDAACXFBH-KFORCWLUNA-N
Synonym 1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate
PubChem CID 12964416
IUPAC Name λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Elemental Analysis 54.7 to 59.3% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 716.48
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis((2R,5R)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less
Product Identifier
  • 1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.