missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%

Product Code. 15465866
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15465866

Brand: Thermo Scientific Alfa Aesar H60592.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147253-67-6 | C18H28P2 | 306.37 g/mol

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147253-67-6
Molecular Formula C18H28P2
Molecular Weight (g/mol) 306.37
MDL Number MFCD00142336
InChI Key AJNZWRKTWQLAJK-KLHDSHLOSA-N
Synonym r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos
PubChem CID 2734551
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C

Specifications

Melting Point 66°C to 76°C
Quantity 1 g
Beilstein 4810602
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 306.36
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.