missing translation for 'onlineSavingsMsg'
Learn More
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%, Thermo Scientific Chemicals Product Code.: 15455436

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%, Thermo Scientific Chemicals

Product Code. 15455436
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15455436

Brand: Thermo Scientific Alfa Aesar H60046.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Sparingly soluble in water.(3.2E-5 g/L) (25°C

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528565-79-9
Molecular Formula C34H36P2
Molecular Weight (g/mol) 506.61
MDL Number MFCD07369027
InChI Key VHHAZLMVLLIMHT-DXCZPEQUNA-N
Synonym --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane,, 1,2-bis 2r,5r-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2beta,5alpha-diphenyltetrahydro-1h-phosphole, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholaneShow MoreShow Less
PubChem CID 11420783
SMILES C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 143°C to 147°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water.(3.2E-5 g/L) (25°C
IUPAC Name (2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Formula Weight 506.6
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-diphenyl-1-phospholanyl]ethane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU30J008 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberH60046.03,H60046.MD
Lot NumberY15I005 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog NumberH60046.03,H60046.MD
Lot NumberX26I001 Certificate TypeCertificate of Analysis Date07/11/2022 Catalog NumberH60046.03,H60046.MD
Lot NumberT19I020 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberH60046.03,H60046.MD
Lot NumberP28I016 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog NumberH60046.03,H60046.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.