missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals Product Code.: 15475916

1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals

Product Code. 15475916
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15475916

Brand: Thermo Scientific Alfa Aesar H60647.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a DuPhos and BPE ligands which are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136735-95-0
Molecular Formula C18H28P2
Molecular Weight (g/mol) 306.37
MDL Number MFCD00142322
InChI Key AJNZWRKTWQLAJK-VGWMRTNUSA-N
Synonym s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane
PubChem CID 2734553
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C

Specifications

Melting Point 66°C to 76°C
Quantity 250 mg
Beilstein 4810602
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 306.36
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM17D025 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberH60647.MD,H60647.03
Lot NumberS10I031 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog NumberH60647.MD,H60647.03
Lot NumberK27Y069 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60647.MD,H60647.03
Lot NumberM17D025 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60647.MD,H60647.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.