missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals Product Code.: 11925961

1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

Product Code. 11925961
500 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 11925961

Brand: Thermo Scientific Acros 435235000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213343-65-8
Molecular Formula C22H40BF4P2Rh
Molecular Weight (g/mol) 556.22
MDL Number MFCD09038435
InChI Key CVUPEPGZCZHSOF-BDNLEJQTNA-N
Synonym 1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborateShow MoreShow Less
PubChem CID 12964417
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 156.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 500 mg
IUPAC Name λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl}-2,5-dimethylphospholane tetrafluoroboranuide
Formula Weight 556.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.