Learn More
1,2-Bis(triethoxysilyl)ethane, 95%, Thermo Scientific Chemicals
£22.61 - £62.31
Chemical Identifiers
CAS | 16068-37-4 |
---|---|
Molecular Formula | C14H34O6Si2 |
Molecular Weight (g/mol) | 354.59 |
MDL Number | MFCD00239362 |
InChI Key | IZRJPHXTEXTLHY-UHFFFAOYSA-N |
Synonym | 1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane |
PubChem CID | 85266 |
IUPAC Name | triethoxy(2-triethoxysilylethyl)silane |
SMILES | CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447008
|
Thermo Scientific Alfa Aesar
H60530.AA |
5 mL |
£22.61
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15457008
|
Thermo Scientific Alfa Aesar
H60530.AC |
25 mL |
£62.31
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.
Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Chemical Identifiers
16068-37-4 | |
354.59 | |
IZRJPHXTEXTLHY-UHFFFAOYSA-N | |
85266 | |
CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC |
C14H34O6Si2 | |
MFCD00239362 | |
1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane | |
triethoxy(2-triethoxysilylethyl)silane |
Specifications
119°C | |
C14H34O6Si2 | |
MFCD00239362 | |
1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane | |
IZRJPHXTEXTLHY-UHFFFAOYSA-N | |
triethoxy(2-triethoxysilylethyl)silane | |
85266 | |
95% | |
0.958 |
16068-37-4 | |
[-CH2Si(OC2H5)3]2 | |
113°C (235°F) | |
Soluble in alcohol,aromatic and aliphatic hydrocarbons. Hydrolyzes with water. | |
CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC | |
354.59 | |
354.59 | |
1.411 | |
1,2-Bis(triethoxysilyl)ethane |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P312-P314-P330-P363-P501c
H301-H312-H372
missing translation for 'einecsNumber' : 240-212-2
missing translation for 'rtecsNumber' : JG7755000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only