missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-2,4-dichlorobenzene, 97+%, Thermo Scientific™

Product Code. 15479047
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 6954-77-4
Molecular Formula C8H7BrCl2O
Molecular Weight (g/mol) 269.95
MDL Number MFCD00043464
InChI Key OEYZGSRLAGSENP-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-2,4-dichlorobenzene, beta-bromo-2,4-dichlorophenetole, b-bromo-2,4-dichlorophenetole, 1-2,4-dichlorophenoxy-2-bromoethane, 1-bromo-2-2,4-dichlorophenoxy ethane, pubchem3151, bromoethoxydichlorobenzene, 2,4-dichlorophenoxyethylbromide, 2,4-dichlorophenoxyethyl bromide, 2-2,4-dichlorophenoxy ethyl bromide
PubChem CID 138881
IUPAC Name 1-(2-bromoethoxy)-2,4-dichlorobenzene
SMILES ClC1=CC=C(OCCBr)C(Cl)=C1

Specifica

Quantity 250 mg
UN Number UN2810
Formula Weight 269.95
Percent Purity ≥97%
Chemical Name or Material 1-(2-Bromoethoxy)-2,4-dichlorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'