Learn More
1-(2-Bromoethyl)naphthalene, 97%, Thermo Scientific Chemicals
POA - £278.00
Chemical Identifiers
CAS | 13686-49-2 |
---|---|
Molecular Formula | C12H11Br |
Molecular Weight (g/mol) | 235.124 |
MDL Number | MFCD00037737 |
InChI Key | GPHCPUFIWQJZOI-UHFFFAOYSA-N |
PubChem CID | 139541 |
IUPAC Name | 1-(2-bromoethyl)naphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11325827
|
Thermo Scientific Alfa Aesar
043491.03 |
1 g |
£81.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11335827
|
Thermo Scientific Alfa Aesar
043491.06 |
5 g |
£278.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11315827
|
Thermo Scientific Alfa Aesar
043491.14 |
25 g | N/A | ||||||
Description
1-(2-Bromoethyl)naphthalene has applications in research field such as Western blotting, Immunoprecipitation, immunogen, and in purification, process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13686-49-2 | |
235.124 | |
GPHCPUFIWQJZOI-UHFFFAOYSA-N | |
1-(2-bromoethyl)naphthalene |
C12H11Br | |
MFCD00037737 | |
139541 | |
C1=CC=C2C(=C1)C=CC=C2CCBr |
Specifications
13686-49-2 | |
>110°C (230°F) | |
MFCD00037737 | |
It is slightly soluble in water. | |
C1=CC=C2C(=C1)C=CC=C2CCBr | |
235.124 | |
235.13 | |
Liquid |
237°C to 238°C (1.65 mmHg) | |
C12H11Br | |
1-2-bromoethyl naphthalene, 2-1-naphthyl ethyl bromide, naphthalene, 1-2-bromoethyl, 2-bromoethylnaphthalene, 1-2-bromoethyl naphtalene, 1-2-bromo-ethyl-naphthalene, naphthalene,1-2-bromoethyl, 2-alpha-naphthyl ethyl bromide, naphthylathylbromid, 2-bromoethyl napthalene | |
GPHCPUFIWQJZOI-UHFFFAOYSA-N | |
1-(2-bromoethyl)naphthalene | |
139541 | |
97% | |
1- (2-Bromoethyl)naphthalene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only