missing translation for 'onlineSavingsMsg'
Learn More
1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals Product Code.: 11304386

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals

Product Code. 11304386
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11304386

Brand: Thermo Scientific Alfa Aesar L20001.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-42-7
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064863,MFCD00674195
InChI Key MUTGBJKUEZFXGO-UHFFFAOYNA-N
Synonym hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydrideShow MoreShow Less
PubChem CID 85689
ChEBI CHEBI:103210
IUPAC Name 3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2CCCCC12

Specifications

Melting Point 32°C to 34°C
Boiling Point 158°C (17 mmHg)
Flash Point 275°C (527°F)
Quantity 500 g
Beilstein 83213
Sensitivity Moisture sensitive
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1,2-Cyclohexanedicarboxylic anhydride, Cis + Trans

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252133 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog NumberL20001.36,L20001.22
Lot NumberU19I002 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog NumberL20001.36,L20001.22
Lot NumberUSLF004235 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL20001.36,L20001.22
Lot Number10160152 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL20001.36,L20001.22
Lot Number10200643 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL20001.36,L20001.22
compliance-icons compliance-icons
Product Identifier
  • 1,2-Cyclohexanedicarboxylic anhydride, cis + trans
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrohexaphthalic anhydride
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.