missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals
£31.20 - £80.40
Chemical Identifiers
CAS | 931-17-9 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00003861 |
InChI Key | PFURGBBHAOXLIO-UHFFFAOYSA-N |
Synonym | 1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans |
PubChem CID | 13601 |
ChEBI | CHEBI:24567 |
IUPAC Name | cyclohexane-1,2-diol |
SMILES | C1CCC(C(C1)O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
931-17-9 | |
116.16 | |
PFURGBBHAOXLIO-UHFFFAOYSA-N | |
13601 | |
cyclohexane-1,2-diol |
C6H12O2 | |
MFCD00003861 | |
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans | |
CHEBI:24567 | |
C1CCC(C(C1)O)O |
Specifications
931-17-9 | |
100.0 | |
White | |
>110°C | |
97.5% min. (GC) | |
C6H10(OH)2 | |
06, 740 | |
Solubility in water: soluble | |
C1CCC(C(C1)O)O | |
116.16 | |
CHEBI:24567 | |
98% | |
1, 2-Cyclohexanediol, 98% |
97.5 | |
73°C to 77°C | |
118°C to 120°C (10.0 mmHg) | |
Authentic | |
C6H12O2 | |
MFCD00003861 | |
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans | |
PFURGBBHAOXLIO-UHFFFAOYSA-N | |
cyclohexane-1,2-diol | |
13601 | |
116.16 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 213-229-8
missing translation for 'rtecsNumber' : GV0220000
missing translation for 'tsca' : TSCA
Product Identifier
- 1,2-Cyclohexanediol, mixture of cis and trans
RUO – Research Use Only