missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1-iodotrifluoroethane, 97%, Thermo Scientific™

Product Code. 11437104
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216394-01-3
Molecular Formula C2Br2F3I
Molecular Weight (g/mol) 367.73
MDL Number MFCD00042521
InChI Key YUBJHLYGOOZDRN-UHFFFAOYNA-N
Synonym 1,2-dibromo-1-iodotrifluoroethane, 1,2-dibromo-2-iodotrifluoroethane, 1,2-dibromo-1,2,2-trifluoro-1-iodoethane, ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
PubChem CID 2736780
IUPAC Name 1,2-dibromo-1,1,2-trifluoro-2-iodoethane
SMILES FC(F)(Br)C(F)(Br)I

Specifications

Density 1.515
Boiling Point 63°C to 65°C (32 mmHg)
Linear Formula BrCFICF2Br
Quantity 1 g
Formula Weight 367.73
Percent Purity 97%
Chemical Name or Material 1,2-Dibromo-1-iodotrifluoroethane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'